2-(2,3-Dihydro-1H-indol-1-yl)-5-fluorophenylamine

Supplier: Matrix Scientific

051220-500MG 051220-2.5G
101894-772EA 364.62 USD
101894-772 101894-774
2-(2,3-Dihydro-1H-indol-1-yl)-5-fluorophenylamine
2-(2,3-Dihydro-1H-indol-1-yl)-5-fluorophenylamine

Hazard: Irritant

Formula: C₁₄H₁₃FN₂
MDL Number: MFCD08699502
CAS Number: 640768-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR