2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-fluoroaniline

Supplier: Matrix Scientific

051219-500MG 051219-2.5G
101894-768EA 364.62 USD
101894-768 101894-770
2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-fluoroaniline
2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-fluoroaniline

Hazard: Irritant

Formula: C₁₅H₁₅FN₂
MDL Number: MFCD08699504
CAS Number: 937597-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR