2-[4-(2-Amino-4-fluorophenyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051218-500MG 051218-2.5G
101894-764EA 364.62 USD
101894-764 101894-766
2-[4-(2-Amino-4-fluorophenyl)-1-piperazinyl]-1-ethanol
2-[4-(2-Amino-4-fluorophenyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₈FN₃O
MDL Number: MFCD12421864
CAS Number: 1155522-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR