2-(2-Ethyl-1-piperidinyl)-5-fluoroaniline
Supplier: Matrix Scientific
051215-500MG
051215-2.5G
101894-752EA
364.62
USD
101894-752
101894-754
2-(2-Ethyl-1-piperidinyl)-5-fluoroaniline
2-(2-Ethyl-1-piperidinyl)-5-fluoroaniline
Hazard: Irritant
Formula:
C₁₃H₁₉FN₂ |
MDL Number:
MFCD11624796 CAS Number: 1095039-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...