4-(2-Amino-4-fluorophenyl)-2-piperazinone

Supplier: Matrix Scientific

051211-500MG 051211-2.5G
101894-736EA 364.62 USD
101894-736 101894-738
4-(2-Amino-4-fluorophenyl)-2-piperazinone
4-(2-Amino-4-fluorophenyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₂FN₃O
MDL Number: MFCD11135640
CAS Number: 1019581-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR