2-(1-Azepanyl)-5-fluoroaniline

Supplier: Matrix Scientific

051210-500MG 051210-2.5G
101894-732EA 364.62 USD
101894-732 101894-734
2-(1-Azepanyl)-5-fluoroaniline
2-(1-Azepanyl)-5-fluoroaniline

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MDL Number: MFCD04971028
CAS Number: 869943-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR