2-(4-Benzyl-1-piperazinyl)-5-fluorophenylamine

Supplier: Matrix Scientific

051209-500MG 051209-2.5G
101894-728EA 364.62 USD
101894-728 101894-730
2-(4-Benzyl-1-piperazinyl)-5-fluorophenylamine
2-(4-Benzyl-1-piperazinyl)-5-fluorophenylamine

Hazard: Irritant

Formula: C₁₇H₂₀FN₃
MDL Number: MFCD08699484
CAS Number: 937597-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR