5-Fluoro-2-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051206-500MG 051206-2.5G
101894-716EA 364.62 USD
101894-716 101894-718
5-Fluoro-2-(2-methyl-1-piperidinyl)aniline
5-Fluoro-2-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MDL Number: MFCD11135623
CAS Number: 952906-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR