1-(2-Amino-4-fluorophenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051202-500MG 051202-2.5G
101894-700EA 364.62 USD
101894-700 101894-702
1-(2-Amino-4-fluorophenyl)-3-pyrrolidinol
1-(2-Amino-4-fluorophenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃FN₂O
MDL Number: MFCD13561456
CAS Number: 1220037-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR