5-Fluoro-2-(1-pyrrolidinyl)aniline
Supplier: Matrix Scientific
051199-500MG
051199-2.5G
101894-688EA
384.62
USD
101894-688
101894-690
5-Fluoro-2-(1-pyrrolidinyl)aniline
5-Fluoro-2-(1-pyrrolidinyl)aniline
Hazard: Irritant
Formula:
C₁₀H₁₃FN₂ |
MDL Number:
MFCD04971020 CAS Number: 869942-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...