2-(2,3-Dihydro-1H-indol-1-yl)-5-methylphenylamine

Supplier: Matrix Scientific

051194-500MG 051194-2.5G
101894-668EA 364.62 USD
101894-668 101894-670
2-(2,3-Dihydro-1H-indol-1-yl)-5-methylphenylamine
2-(2,3-Dihydro-1H-indol-1-yl)-5-methylphenylamine

Hazard: Irritant

Formula: C₁₅H₁₆N₂
MDL Number: MFCD11645445
CAS Number: 1097786-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR