2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-methylaniline

Supplier: Matrix Scientific

051193-500MG 051193-2.5G
101894-664EA 364.62 USD
101894-664 101894-666
2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-methylaniline
2-[3,4-Dihydro-2(1H)-isoquinolinyl]-5-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₈N₂
MDL Number: MFCD11547642
CAS Number: 1097786-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR