2-[4-(2-Amino-4-methylphenyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051192-500MG 051192-2.5G
101894-660EA 364.62 USD
101894-660 101894-662
2-[4-(2-Amino-4-methylphenyl)-1-piperazinyl]-1-ethanol
2-[4-(2-Amino-4-methylphenyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₃H₂₁N₃O
MDL Number: MFCD12421861
CAS Number: 1155570-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR