1-(2-Amino-4-methylphenyl)-4-piperidinol

Supplier: Matrix Scientific

051186-500MG 051186-2.5G
101894-636EA 461.54 USD
101894-636 101894-638
1-(2-Amino-4-methylphenyl)-4-piperidinol
1-(2-Amino-4-methylphenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD11645436
CAS Number: 1097785-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR