2-(4-Benzyl-1-piperazinyl)-5-methylphenylamine

Supplier: Matrix Scientific

051183-500MG 051183-2.5G
101894-624EA 364.62 USD
101894-624 101894-626
2-(4-Benzyl-1-piperazinyl)-5-methylphenylamine
2-(4-Benzyl-1-piperazinyl)-5-methylphenylamine

Hazard: Irritant

Formula: C₁₈H₂₃N₃
MDL Number: MFCD13561454
CAS Number: 946719-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR