5-Methyl-2-(4-methyl-1-piperazinyl)aniline

Supplier: Matrix Scientific

051182-500MG 051182-2.5G
101894-620EA 364.62 USD
101894-620 101894-622
5-Methyl-2-(4-methyl-1-piperazinyl)aniline
5-Methyl-2-(4-methyl-1-piperazinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD10686569
CAS Number: 946731-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR