5-Methyl-2-(3-methyl-1-piperidinyl)aniline
Supplier: Matrix Scientific
051180-500MG
051180-2.5G
101894-612EA
364.62
USD
101894-612
101894-614
5-Methyl-2-(3-methyl-1-piperidinyl)aniline
5-Methyl-2-(3-methyl-1-piperidinyl)aniline
Hazard: Irritant
Formula:
C₁₃H₂₀N₂ |
MDL Number:
MFCD11643967 CAS Number: 946684-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...