2-[2-Amino(ethyl)-4-methylanilino]-1-ethanol

Supplier: Matrix Scientific

051177-500MG 051177-2.5G
101894-600EA 364.62 USD
101894-600 101894-602
2-[2-Amino(ethyl)-4-methylanilino]-1-ethanol
2-[2-Amino(ethyl)-4-methylanilino]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD12813174
CAS Number: 1183355-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR