1-(2-Amino-4-methylphenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051175-500MG 051175-2.5G
101894-592EA 364.62 USD
101894-592 101894-594
1-(2-Amino-4-methylphenyl)-3-pyrrolidinol
1-(2-Amino-4-methylphenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD13561452
CAS Number: 1220034-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR