5-Methyl-2-(4-morpholinyl)aniline

Supplier: Matrix Scientific

051174-500MG 051174-2.5G
101894-588EA 364.62 USD
101894-588 101894-590
5-Methyl-2-(4-morpholinyl)aniline
5-Methyl-2-(4-morpholinyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
MDL Number: MFCD09755879
CAS Number: 91429-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR