2-[3,4-Dihydro-1(2H)-quinolinyl]-5-(trifluoromethyl)aniline

Supplier: Matrix Scientific

051166-500MG 051166-2.5G
101894-556EA 364.62 USD
101894-556 101894-558
2-[3,4-Dihydro-1(2H)-quinolinyl]-5-(trifluoromethyl)aniline
2-[3,4-Dihydro-1(2H)-quinolinyl]-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₅F₃N₂
MDL Number: MFCD08699428
CAS Number: 937596-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR