2-{4-[2-Amino-4-(trifluoromethyl)phenyl]-1-piperazinyl}-1-ethanol

Supplier: Matrix Scientific

051163-500MG 051163-2.5G
101894-544EA 730.77 USD
101894-544 101894-546
2-{4-[2-Amino-4-(trifluoromethyl)phenyl]-1-piperazinyl}-1-ethanol
2-{4-[2-Amino-4-(trifluoromethyl)phenyl]-1-piperazinyl}-1-ethanol

Hazard: Irritant

Formula: C₁₃H₁₈F₃N₃O
MDL Number: MFCD12676535
CAS Number: 1155523-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR