4-[2-Amino-4-(trifluoromethyl)phenyl]-2-piperazinone
Supplier: Matrix Scientific
051156-500MG
051156-2.5G
101894-516EA
364.62
USD
101894-516
101894-518
4-[2-Amino-4-(trifluoromethyl)phenyl]-2-piperazinone
4-[2-Amino-4-(trifluoromethyl)phenyl]-2-piperazinone
Hazard: Irritant
Formula:
C₁₁H₁₂F₃N₃O |
MDL Number:
MFCD09046843 CAS Number: 882672-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...