2-(1-Azepanyl)-5-(trifluoromethyl)aniline

Supplier: Matrix Scientific

051155-500MG 051155-2.5G
101894-512EA 364.62 USD
101894-512 101894-514
2-(1-Azepanyl)-5-(trifluoromethyl)aniline
2-(1-Azepanyl)-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₇F₃N₂
MW: 258.29 g/mol
MDL Number: MFCD08246126
CAS Number: 858126-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR