2-(4-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

051153-500MG 051153-2.5G
101894-504EA 364.62 USD
101894-504 101894-506
2-(4-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine
2-(4-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₇F₃N₂
MDL Number: MFCD03034595
CAS Number: 445465-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR