2-(3-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

051152-500MG 051152-2.5G
101894-500EA 615.38 USD
101894-500 101894-502
2-(3-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine
2-(3-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₇F₃N₂
MDL Number: MFCD08699404
CAS Number: 926208-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR