2-(2-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

051151-500MG 051151-2.5G
101894-496EA 615.38 USD
101894-496 101894-498
2-(2-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine
2-(2-Methyl-1-piperidinyl)-5-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₇F₃N₂
MDL Number: MFCD09047962
CAS Number: 926272-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR