2-[2-Amino(methyl)-4-(trifluoromethyl)anilino]-1-ethanol

Supplier: Matrix Scientific

051146-500MG 051146-2.5G
101894-476EA 364.62 USD
101894-476 101894-478
2-[2-Amino(methyl)-4-(trifluoromethyl)anilino]-1-ethanol
2-[2-Amino(methyl)-4-(trifluoromethyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₃F₃N₂O
MDL Number: MFCD11642967
CAS Number: 1098343-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR