5-Chloro-2-(2,3-dihydro-1H-indol-1-yl)aniline

Supplier: Matrix Scientific

051139-500MG 051139-2.5G
101894-448EA 364.62 USD
101894-448 101894-450
5-Chloro-2-(2,3-dihydro-1H-indol-1-yl)aniline
5-Chloro-2-(2,3-dihydro-1H-indol-1-yl)aniline

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂
MDL Number: MFCD08696896
CAS Number: 937606-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR