5-Chloro-2-[3,4-dihydro-2(1H)isoquinolinyl]aniline

Supplier: Matrix Scientific

051138-500MG 051138-2.5G
101894-444EA 364.62 USD
101894-444 101894-446
5-Chloro-2-[3,4-dihydro-2(1H)isoquinolinyl]aniline
5-Chloro-2-[3,4-dihydro-2(1H)isoquinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂
MW: 258.75 g/mol
MDL Number: MFCD07365175
CAS Number: 893751-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR