5-Chloro-2-(2-ethyl-1-piperidinyl)aniline
Supplier: Matrix Scientific
051134-500MG
051134-2.5G
101894-428EA
423.08
USD
101894-428
101894-430
5-Chloro-2-(2-ethyl-1-piperidinyl)aniline
5-Chloro-2-(2-ethyl-1-piperidinyl)aniline
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂ MW: 238.76 g/mol |
MDL Number:
MFCD11618643 CAS Number: 946669-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...