N-(2-Amino-4-chlorophenyl)-N-cyclohexyl-N-methylamine

Supplier: Matrix Scientific

051133-500MG 051133-2.5G
101894-424EA 364.62 USD
101894-424 101894-426
N-(2-Amino-4-chlorophenyl)-N-cyclohexyl-N-methylamine
N-(2-Amino-4-chlorophenyl)-N-cyclohexyl-N-methylamine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD07365178
CAS Number: 893751-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR