4-(2-Amino-4-chlorophenyl)-2-piperazinone
Supplier: Matrix Scientific
051130-500MG
051130-2.5G
101894-412EA
364.62
USD
101894-412
101894-414
4-(2-Amino-4-chlorophenyl)-2-piperazinone
4-(2-Amino-4-chlorophenyl)-2-piperazinone
Hazard: Irritant
Formula:
C₁₀H₁₂ClN₃O |
MDL Number:
MFCD09049174 CAS Number: 926239-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...