2-(1-Azepanyl)-5-chloroaniline

Supplier: Matrix Scientific

051129-500MG 051129-2.5G
101894-408EA 364.62 USD
101894-408 101894-410
2-(1-Azepanyl)-5-chloroaniline
2-(1-Azepanyl)-5-chloroaniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD04000824
CAS Number: 710301-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR