2-(4-Benzyl-1-piperazinyl)-5-chlorophenylamine

Supplier: Matrix Scientific

051128-500MG 051128-2.5G
101894-404EA 364.62 USD
101894-404 101894-406
2-(4-Benzyl-1-piperazinyl)-5-chlorophenylamine
2-(4-Benzyl-1-piperazinyl)-5-chlorophenylamine

Hazard: Irritant

Formula: C₁₇H₂₀ClN₃
MW: 301.82 g/mol
MDL Number: MFCD07365174
CAS Number: 893751-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR