1-(2-Amino-4-chlorophenyl)-3-pyrrolidinol
Supplier: Matrix Scientific
051121-500MG
051121-2.5G
101894-376EA
364.62
USD
101894-376
101894-378
1-(2-Amino-4-chlorophenyl)-3-pyrrolidinol
1-(2-Amino-4-chlorophenyl)-3-pyrrolidinol
Hazard: Irritant
Formula:
C₁₀H₁₃ClN₂O |
MDL Number:
MFCD13561446 CAS Number: 1220037-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...