3-Chloro-2-[3,4-dihydro-1(2H)-quinolinyl]aniline

Supplier: Matrix Scientific

051114-500MG 051114-2.5G
101894-348EA 364.62 USD
101894-348 101894-350
3-Chloro-2-[3,4-dihydro-1(2H)-quinolinyl]aniline
3-Chloro-2-[3,4-dihydro-1(2H)-quinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂
MDL Number: MFCD08700275
CAS Number: 937604-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR