3-Chloro-2-(2,3-dihydro-1H-indol-1-yl)aniline
Supplier: Matrix Scientific
051113-500MG
051113-2.5G
101894-344EA
364.62
USD
101894-344
101894-346
3-Chloro-2-(2,3-dihydro-1H-indol-1-yl)aniline
3-Chloro-2-(2,3-dihydro-1H-indol-1-yl)aniline
Hazard: Irritant
Formula:
C₁₄H₁₃ClN₂ |
MDL Number:
MFCD08700274 CAS Number: 937604-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...