2-[4-(2-Amino-6-chlorophenyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051111-500MG 051111-2.5G
101894-336EA 364.62 USD
101894-336 101894-338
2-[4-(2-Amino-6-chlorophenyl)-1-piperazinyl]-1-ethanol
2-[4-(2-Amino-6-chlorophenyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃O
MDL Number: MFCD12421866
CAS Number: 1155523-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR