N-(2-Amino-6-chlorophenyl)-N-cyclohexyl-N-ethylamine

Supplier: Matrix Scientific

051110-500MG 051110-2.5G
101894-332EA 364.62 USD
101894-332 101894-334
N-(2-Amino-6-chlorophenyl)-N-cyclohexyl-N-ethylamine
N-(2-Amino-6-chlorophenyl)-N-cyclohexyl-N-ethylamine

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂
MDL Number: MFCD11126051
CAS Number: 1039833-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR