3-Chloro-2-(2-ethyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051108-500MG 051108-2.5G
101894-324EA 364.62 USD
101894-324 101894-326
3-Chloro-2-(2-ethyl-1-piperidinyl)aniline
3-Chloro-2-(2-ethyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD11618641
CAS Number: 946730-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR