N-(2-Amino-6-chlorophenyl)-N-cyclohexyl-N-methylamine

Supplier: Matrix Scientific

051107-500MG 051107-2.5G
101894-320EA 364.62 USD
101894-320 101894-322
N-(2-Amino-6-chlorophenyl)-N-cyclohexyl-N-methylamine
N-(2-Amino-6-chlorophenyl)-N-cyclohexyl-N-methylamine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD11156648
CAS Number: 1021238-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR