1-(2-Amino-6-chlorophenyl)-4-piperidinol

Supplier: Matrix Scientific

051105-500MG 051105-2.5G
101894-312EA 364.62 USD
101894-312 101894-314
1-(2-Amino-6-chlorophenyl)-4-piperidinol
1-(2-Amino-6-chlorophenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD09739176
CAS Number: 954271-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR