4-(2-Amino-6-chlorophenyl)-2-piperazinone
Supplier: Matrix Scientific
051104-500MG
051104-2.5G
101894-308EA
364.62
USD
101894-308
101894-310
4-(2-Amino-6-chlorophenyl)-2-piperazinone
4-(2-Amino-6-chlorophenyl)-2-piperazinone
Hazard: Irritant
Formula:
C₁₀H₁₂ClN₃O |
MDL Number:
MFCD09043790 CAS Number: 926260-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...