3-Chloro-2-(3-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051101-500MG 051101-2.5G
101894-296EA 364.62 USD
101894-296 101894-298
3-Chloro-2-(3-methyl-1-piperidinyl)aniline
3-Chloro-2-(3-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD08691866
CAS Number: 915921-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR