3-Chloro-2-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051100-500MG 051100-2.5G
101894-292EA 364.62 USD
101894-292 101894-294
3-Chloro-2-(2-methyl-1-piperidinyl)aniline
3-Chloro-2-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD09043188
CAS Number: 926215-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR