1-(2-Amino-6-chlorophenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051096-500MG 051096-2.5G
101894-276EA 364.62 USD
101894-276 101894-278
1-(2-Amino-6-chlorophenyl)-3-pyrrolidinol
1-(2-Amino-6-chlorophenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD13561443
CAS Number: 1220037-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR