2-(2-Ethyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051085-500MG 051085-2.5G
101894-232EA 364.62 USD
101894-232 101894-234
2-(2-Ethyl-1-piperidinyl)aniline
2-(2-Ethyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD11618638
CAS Number: 946730-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR