2-(2-Methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051079-500MG 051079-2.5G
101894-208EA 186.15 USD
101894-208 101894-210
2-(2-Methyl-1-piperidinyl)aniline
2-(2-Methyl-1-piperidinyl)aniline

Hazard: Irritant/Refrigerate

Formula: C₁₂H₁₈N₂
Melting Pt: 44…45 °C
MDL Number: MFCD04970972
CAS Number: 67150-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR