2-[2-Amino(ethyl)anilino]-1-ethanol

Supplier: Matrix Scientific

051077-500MG 051077-2.5G
101894-200EA 364.62 USD
101894-200 101894-202
2-[2-Amino(ethyl)anilino]-1-ethanol
2-[2-Amino(ethyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD12813169
CAS Number: 22271-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR